Learn More
Trajan™ SilTite™ FingerTite Blanking Ferrule
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand:  Trajan™ 073633
252.18 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15248983
  • 259.00 DKK / Pack of 2
Only null left
Estimated Shipment: 09-10-2025
Description
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
Blank Ferrule
SilTite
2/Pk.