Learn More
Epredia™ Double Cytofunnel™
Shop All Epredia Products
Click to view available options
Quantity:
25 Pk.
500/Cs.
Unit Size:
Pack of 25
Pack of 500
Description
- Produces more cost-effective, thin-layer preparations when compared to other thin-layer methods
- Reduces the risk of exposure to pathogenic samples
- Reduces preparation time
- Disposable to eliminate time-consuming decontamination and cleaning
- Epredia™ Double Cytofunnel™ REQUIRES Epredia™ Cytoclips™
- Decreases the possibility of cross-contamination
- Features funnel markings corresponding to slide markings for simple identification
Includes:
- Pre-attached filter card
- Closure caps
- Easy-to-use operational illustrations
Specifications
Specifications
Color | White |
Disposable | Single-use |
Quantity | 25 Pk. |
Type | Double Cytofunnel™, With Filter Cards and Caps |
Volume (Metric) | 0.05 to 0.25 mL |