Learn More
Epredia™ Slimsette™ Biopsy Cassettes in Tube Packs, aqua
Aid the process of speedy and reliable biopsy specimen labeling with Epredia™ Slimsette™ Biopsy Cassettes in Tube Packs, for use with automatic labeling instruments.
1981.73 DKK
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12668316
|
Epredia™
B1000737AQ |
Aqua | N/A | ||||||
12678316
|
Epredia™
B1000737BL |
Blue | N/A | ||||||
12688316
|
Epredia™
B1000737GR |
Green | N/A | ||||||
12698316
|
Epredia™
B1000737GY |
Gray | N/A | ||||||
12608326
|
Epredia™
B1000737LI |
Lilac | N/A | ||||||
12618326
|
Epredia™
B1000737OR |
Orange | N/A | ||||||
12628326
|
Epredia™
B1000737PE |
Peach | N/A | ||||||
12638326
|
Epredia™
B1000737PK |
Pink | N/A | ||||||
12648326
|
Epredia™
B1000737TN |
Tan | N/A | ||||||
12658326
|
Epredia™
B1000737WH |
White | N/A | ||||||
12678326
|
Epredia™
B1000737YW |
Yellow | N/A | ||||||
Description
Specifications
Small Square Pore | |
Biopsy Cassette |