Learn More
Epredia™ Slimsette™ Embedding Cassettes in Tube Packs, aqua
Aid the process of speedy and reliable specimen labeling with these handily packaged cassettes, designed for use with automatic labeling instruments.
1975.77 DKK
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12668306
|
Epredia™
B1000736AQ |
Aqua | |||||||
12678306
|
Epredia™
B1000736BL |
Blue | |||||||
12688306
|
Epredia™
B1000736GR |
Green | |||||||
12698306
|
Epredia™
B1000736GY |
Gray | |||||||
10259788
|
Epredia™
B1000736LI |
Lilac | |||||||
12608316
|
Epredia™
B1000736OR |
Orange | |||||||
12618316
|
Epredia™
B1000736PE |
Peach | |||||||
12628316
|
Epredia™
B1000736PK |
Pink | |||||||
12638316
|
Epredia™
B1000736TN |
Tan | |||||||
12658316
|
Epredia™
B1000736YW |
Yellow | |||||||