Learn More
Sodium Hydroxide Solution, 0.111M (0.111N), for analysis of milk, Fisher Chemical™
Volumetric sodium hydroxide solution suitable for the determination of the acidity of milk.
40.00€ - 117.00€
Specifications
Normality | 0.111 |
---|---|
Packaging | HDPE Plastic Bottle |
Traceability to NIST | Traceable to NIST, used to titrate the lactic acid present in milk |
Physical Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958776
|
Fisher Chemical
J/7670/15 |
1 L |
40.00€
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15968776
|
Fisher Chemical
J/7670/21 |
5 L | N/A | N/A | N/A | ||||
Description
Specifications
0.111 | |
HDPE Plastic Bottle | |
Traceable to NIST, used to titrate the lactic acid present in milk | |
Liquid |