Learn More
MAPA™ SOLO 992 Disposable Latex Gloves
These single use gloves are ideal for general use in hospitals, labs and clinics as well as drug manufacturing and pharmaceutical industries, mechanical/automotive industries and manipulation of precision parts.
Specifications
Certifications/Compliance | PPE Cat III, EN ISO 374-1 Type C, EN ISO 374-5 |
---|---|
Content And Storage | Keep gloves in their packaging away from light, moisture, and heat sources |
Description | Not recommended for use with dithiocarbamates and thiazoles, or for individuals with latex sensitivity or allergy |
Color | Natural |
Disposable | Single-use |
Description
- PPE Category III
- Single use only
- Natural latex material
- Wrist rolled edge
- Natural color
- Powder-coated interior finish
- Smooth exterior finish
- Guaranteed silicone-free
- Excellent dexterity due to latex flexibility
- Easy donning and doffing
Specifications
PPE Cat III, EN ISO 374-1 Type C, EN ISO 374-5 | |
Not recommended for use with dithiocarbamates and thiazoles, or for individuals with latex sensitivity or allergy | |
Single-use | |
No | |
0.10 mm |
Keep gloves in their packaging away from light, moisture, and heat sources | |
Natural | |
Laboratory | |
Latex (Silicone-free) | |
24 cm |