Learn More
Fisherbrand™ Solutions A+ Aloe Latex Gloves
Protects hands from drying, chapping and cracking. Fisherbrand™ Solutions A+ Aloe Latex Gloves combine protection and comfort.
Specifications
Certifications/Compliance | ISO 9001 QMS, FDA Biocompatibility Guidance, Passes Viral Penetration Test (ASTM F1671), 510K Approved |
---|---|
Color | Natural |
Hand | Ambidextrous |
Material | Latex |
Powdered | Powder Free |
Description
Latex gloves provide a protective barrier and contain soothing Aloe vera to help prevent skin from drying, cracking and chapping. Textured to provide excellent grip. Powder free. EN 374:2003 (AQL 1.5).- PPE Category III
- Disposable
- Aloe vera coating protects from drying, chapping and cracking
- Ideal for prolonged use
- Textured surface provides good grip
- Latex
Specifications
ISO 9001 QMS, FDA Biocompatibility Guidance, Passes Viral Penetration Test (ASTM F1671), 510K Approved | |
Ambidextrous | |
Powder Free | |
5.5 mil | |
Beaded/Rolled | |
240 mm |
Natural | |
Latex | |
Non-sterile | |
Ideal for workers who have frequent glove changes | |
18 MPa |