Learn More
Sampling Systems™ Stainless Steel Bailers
Suitable for use in Pharmaceutical, Chemical and Hygienic applications. Sampling Systems™ Stainless Steel Bailers have a fully welded handle to give a hygienic, crevice free finish.
Brand:  Sampling Systems™ A296-2
670.39 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12795008
  • 738.00 DKK / Each
Only null left
Estimated Shipment: 10-10-2025
Description
  • Crevice free
  • Satin finish
  • Flat cut rim on bowl
  • 316L Stainless Steel
Specifications
Specifications
Bailer (Heavy-duty)
2 L
340 mm
Stainless Steel (316L)
210 mm