Learn More
Grant Instruments™ Stainless Steel Bath Cover
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Bath Cover for Grant Stirred Water Baths is available flat or gabled with removable hinge.
Brand:  Grant Instruments™ STL5
783.40 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12787779
  • 864.24 DKK / Each
Only null left
Estimated Shipment: 07-10-2025
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Bath Cover for Grant Stirred Water Baths is available flat or gabled with removable hinge.
  • Stainless steel construction
  • Plastic handle
  • Available flat (STL 5), or gabled with removable hinge (STL 12, STL 38)
  • For use with Grant Stirred Water Baths and Heated Circulators
  • Prevents heat from escaping
  • Reduces sample contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ST5 tank