Learn More
Memmert™ Stainless Steel Bottom Drip Tray for SN110plus, 15 mm
Non-perforated bottom drip trays constructed of stainless steel.
Brand:  Memmert™ SN110plusB04359
1439.97 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17558455
  • 1480.00 DKK / Each
Only null left
Estimated Shipment: 02-10-2025
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SN110plus Sterilizer
Bottom Drip Tray
Rim: 15 mm