Learn More
Tenak™ Stainless Steel Freezer Rack, 130 mm Box Height
Stainless Steel Freezer Rack is designed for use with freezers.
770.10 DKK - 1010.00 DKK
Specifications
Holds Boxes
Material Stainless Steel
For Use With (Equipment) Freezers
Height (Metric) 130 mm
View More Specs
Products 5
Product Code Brand Positions Price Quantity & Availability  
Product Code Brand Positions Price Quantity & Availability  
12425950
View Documents
Tenak™
TE25180
6 N/A
View alternative products
Save Up to 
»
N/A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
12435950
View Documents
Tenak™
TE25182
8 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12445950
View Documents
Tenak™
TE25184
12 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12455950
View Documents
Tenak™
TE25186
10 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12465950
View Documents
Tenak™
TE25188
15 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • For use with freezers
  • Constructed of stainless steel
Specifications
Specifications
Boxes
Stainless Steel
Freezers
130 mm