Learn More
Grant Instruments™ Stainless Steel Lid
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Lid for Grant Stirred Water Baths is for continuous continuous use with water above 90°C.
Brand: Grant Instruments™ LST12
1932.39 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Lid for Grant Stirred Water Baths is for continuous continuous use with water above 90°C.
- Stainless steel construction
- Gabled with plastic handle
- For use with Grant Stirred Water Baths and Heated Circulators
- Prevents heat from escaping
- Reduces sample contamination
- For continuous use with water above 90°C.
Specifications
Length: 196 | |
ST12 tank | |
Stainless Steel |