Learn More
Grant Instruments™ Stainless Steel Lid
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Lid for Grant Stirred Water Baths is for continuous continuous use with water above 90°C.
Brand:  Grant Instruments™ LST26
2917.24 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15162625
  • 2995.00 DKK / Each
Only null left
Estimated Shipment: 29-09-2025
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Lid for Grant Stirred Water Baths is for continuous continuous use with water above 90°C.
  • Stainless steel construction
  • Gabled with plastic handle
  • For use with Grant Stirred Water Baths and Heated Circulators
  • Prevents heat from escaping
  • Reduces sample contamination
  • For continuous use with water above 90°C.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Length: 375
ST18, ST26 tanks
90°C