Learn More
Sartorius™ Stainless Steel OIML Class F1 Reference Line Weight Sets Without DAkkS Calibration Certificate
POA - 32735.00 DKK
Specifications
Case Material Wood
View More Specs
Products 6
Product Code Brand Mass Price Quantity & Availability  
Product Code Brand Mass Price Quantity & Availability  
10564021
View Documents
Sartorius™
YCS31-613-00
1 g to 1 kg
14155.00 DKK
1 set
Only null left
Estimated Shipment: 18-08-2025
Add to basket
10195640
View Documents
Sartorius™
YCS31-713-00
1 g to 10 kg
POA
1 set
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
10094240
View Documents
Sartorius™
YCS01-513-00
1 mg to 100 g
9795.00 DKK
1 set
Only null left
Estimated Shipment: 21-07-2025
Add to basket
10145550
View Documents
Sartorius™
YCS01-613-00
1 mg to 1 kg
17135.00 DKK
1 set
Only null left
Estimated Shipment: 18-08-2025
Add to basket
10155550
View Documents
Sartorius™
YCS01-653-00
1 mg to 5 kg
32735.00 DKK
1 set
Only null left
Estimated Shipment: 18-08-2025
Add to basket
10004250
View Documents
Sartorius™
YCS01-523-00
1 mg to 200 g
11935.00 DKK
1 set
Only null left
Estimated Shipment: 21-07-2025
Specifications
Wood