Learn More
Fisherbrand™ Scaffolding Feet
Provides a stable base for laboratory scaffolding. Fisherbrand™ Scaffolding Feet come with holes for screwing the foot to the bench, floor or wall.
Brand:  Fisherbrand™ 2117/1
1254.45 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11588112
  • 1260.00 DKK / Pack of 4
Only null left
Estimated Shipment: 03-11-2025
Add to basket
Description
Description
  • Allen key provided
  • Socket head screw fixing for rods or tubes of 13mm diameter
  • Manufactured from either stainless steel or aluminum
Specifications
Specifications
Stainless Steel
Stand Foot