Learn More
Tenak™ Standard Rack for Upright Freezer, 130 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
774.00 DKK - 990.00 DKK
Specifications
Holds | Boxes |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 130 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12419530
|
Tenak™
TE23334 |
Boxes |
892.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
12439530
|
Tenak™
TE23336 |
Boxes |
892.00 DKK
Each |
Estimated Shipment: 12-08-2025
Log in to see stock. |
|||||
12459520
|
Tenak™
TE23328 |
Boxes |
774.00 DKK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
12459530
|
Tenak™
TE23338 |
Boxes |
990.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
12479520
|
Tenak™
TE23330 |
Boxes |
831.00 DKK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
12499520
|
Tenak™
TE23332 |
Boxes |
831.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
Description
- Measure the inside height and depth of your freezer when choosing the right rack
- Racks can be used with all freezer brands
- Constructed of stainless steel
Specifications
Boxes | |
Stainless Steel | |
Upright Freezers | |
130 mm (Box) |