Learn More
Bel-Art™ Sterileware™ Extra-Long, Bent Handle Spoons
Ideal for sampling and dispensing of powders, granules, and liquids.
3243.16 DKK - 3545.00 DKK
Specifications
Product Type | Spoon |
---|---|
Material | HDPE |
Autoclavable | Not Autoclavable |
Quantity | 100/Pk. |
Length (Metric) Handle | 30 cm |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
16856916
|
Bel-Art™
F36947-0010 |
10 mL |
3545.00 DKK
Pack of 100 |
Estimated Shipment: 15-08-2025
Log in to see stock. |
|||||
16866916
|
Bel-Art™
F36947-0020 |
20 mL |
3243.16 DKK
Pack of 100 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
Description
- Long handle allows sampling from a distance reducing the risk of contamination
- Handle is bent at bowl to facilitate easier retrieval of samples, including from within tall containers
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) spoons heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Spoon | |
Not Autoclavable | |
30 cm |
HDPE | |
100/Pk. |