Learn More
IBA Lifesciences Streptamer™ CD3/CD28 Kit for T Cell Expansion
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
3 x 160 μL
Unit Size:
1 set
Description
- CD3 Fab-Strep for T cell expansion and CD28 Fab-Strep for T cell expansion should be stored at -90 to -60°C.
- Strep-Tactin™ multimer for T cell expansion should be stored at 2 to 8°C.
- No component is classified as hazardous according to (EC) No 1272/2008 [CLP].
- The kit is good for stimulation of 1.6 x 107 cells in total.
- After the mixing of the single reagents, thereby producing the activating complex, the mixture is stable for 3 months at 2 to 8°C without losing its functionality.
- All components are sterile filtered and do not contain any preservatives.
- Please handle under sterile conditions.
- They are non-magnetic soluble protein complexes generated by multimerization of αCD3- and αCD28- Fab-Streps with a Strep-Tactin™ multimer
- They are completely reversible reagents; i.e.
- they can be removed from the cells by the addition of biotin and hence the extent of stimulation/expansion can be precisely controlled.
Specifications
Specifications
Capacity (Metric) | 3 x 160 μL |
Kit Contents | 160 μL CD3 Fab-Strep, 160 μL CD28 Fab-Strep, 160 μL Strep-Tactin™ multimer |
Quantity | 3 x 160 μL |
For Use With (Application) | T Cell Expansion of 1.6 x 107 Cells |