Learn More
TCR Beta Armenian Hamster anti-Mouse, PE, Clone: H57-597, Proteintech
Armenian Hamster Monoclonal Antibody
Marque: Proteintech PE-65106-100UG
Description
The TCR Beta antibody from Proteintech is a armenian hamster monoclonal antibody generated with affinity purified tcr from mouse do-11.10 cells of mouse TCR Beta. This antibody recognizes mouse antigen. The TCR Beta antibody has been validated for the following applications: FC analysis. This antibody is pe.
Spécification
TCR Beta | |
Monoclonal | |
0.2 mg/mL | |
Armenian Hamster | |
Primary | |
Mouse |
Flow Cytometry | |
H57-597 | |
PE | |
100 μg | |
21577 | |
IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.