Learn More
RETSCH Teflon Grinding Jars
2400.00 DKK - 4785.00 DKK
Product Code | Brand | For Use With (Equipment) | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Capacity (Metric) | Price | Quantity & Availability | ||||
10068931
|
RETSCH
02.462.0184 |
For MM 200 | 10 mL |
2400.00 DKK
Each |
Estimated Shipment: 23-10-2025
Log in to see stock. |
||||
10054290
|
RETSCH
02.462.0051 |
For MM 200 | 25 mL |
4785.00 DKK
Each |
Estimated Shipment: 23-09-2025
Log in to see stock. |
||||
Specifications
Grinding Jar | |
Teflon |