Learn More
Terbium Standard for ICP/MS - Claritas PPT Grade, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
Specifications
Specifications
CAS | 7440-27-9 |
Molecular Formula | Tb |
MDL Number | MFCD00011256 |
InChI Key | GZCRRIHWUXGPOV-UHFFFAOYSA-N |
SMILES | [Tb] |
IUPAC Name | terbium |
Solvent or Matrix | 2% HNO3 |
Molecular Weight (g/mol) | 158.93 |
PubChem CID | 23958 |
Quantity | 125 mL |