Learn More
SCAT™ PTFE Thread Adapters
SCAT™ Safety Caps and SCAT™ Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
Brand: SCAT™ 107632
231.73 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
SCAT™ thread adapters are ideally suited for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polytetrafluoroethylene (PTFE) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Specifications
UNF 1/4 in. 28 G (f) to GL14 (m) | |
PTFE | |
Thread Adapter |