Learn More
Sakura Finetek™ Tissue-Tek™ O.C.T. Compound
This optimum cutting temperature (O.C.T.) formulation of water-soluble glycols and resins provides a convenient specimen matrix for cryostat sectioning at temperatures of -10°C and below.
Brand: Sakura Finetek™ 4583
Description
- Leaves no residue on slides during staining procedure
- Eliminates undesirable background staining
- Provided in a 118mL bottle
Specifications
O.C.T. Compound | |
Staining procedures | |
118 mL Bottle |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.