Learn More
Thermo Scientific™ Titanium Buckets for SureSpin™ 632 rotor
For use with SureSpin™ 632 rotor
150715.14 DKK - 155214.09 DKK
Specifications
For Use With (Equipment) | SureSpin 632 |
---|---|
Material | Titanium |
Quantity | 6/Pk. |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
16634884
|
Thermo Scientific™
75003036 |
17 mL |
150715.14 DKK
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16644884
|
Thermo Scientific™
75003037 |
38.5 mL |
155214.09 DKK
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
SureSpin 632 | |
Titanium | |
6/Pk. |