Learn More
SI Analytics™ Titration Vessel
Temperature controlled with double jacketed
Brand: SI Analytics™ 285216070
5103.30 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Glass vessel
- Double jacket
Specifications
30 to 150 mL | |
Tritation vessel | |
Titration | |
Glass |