missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transketolase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11039-1-AP-20UL
This item is not returnable.
View return policy
Description
Transketolase encodes a thiamine-dependent enzyme which plays a role in the channeling of excess sugar phosphates to glycolysis in the pentose phosphate pathway. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
Transketolase | |
Polyclonal | |
Unconjugated | |
TKT | |
TK, TKT, TKT1, transketolase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21881, 64524, 7086 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P29401, P40142, P50137 | |
Tkt | |
Transketolase Fusion Protein Ag1519 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction