Learn More
Trimethyl-p-tolylsilane, 95%, Thermo Scientific Chemicals
Product Code. 11403498
Click to view available options
Quantity:
5 g
25 g
It is employed in the production of trimethyl-(4-trimethylsilanyl-benzyl)-silane by reacting with chloro-trimethyl-silane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the production of trimethyl-(4-trimethylsilanyl-benzyl)-silane by reacting with chloro-trimethyl-silane.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3728-43-6
Molecular Formula C10H16Si
Molecular Weight (g/mol) 164.323
MDL Number MFCD00014850
InChI Key QGHURGPPCGMAMZ-UHFFFAOYSA-N
Synonym trimethyl-p-tolylsilane, p-tolyltrimethylsilane, trimethyl 4-methylphenyl silane, trimethyl-p-tolyl silane, trimethyl p-tolyl silane, 4-trimethylsilyl toluene, silane, p-tolyl-trimethyl-,, 4-methylphenyl trimethylsilane
PubChem CID 298149
IUPAC Name trimethyl-(4-methylphenyl)silane
SMILES CC1=CC=C(C=C1)[Si](C)(C)C
Specifications
Boiling Point 191°C to 192°C
Quantity 25 g
UN Number UN1993
Flash Point 46°C (115°F)
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 164.33
Percent Purity 97%
Refractive Index 1.491
Density 0.865
Chemical Name or Material Trimethyl-p-tolylsilane