missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBGCP2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
Antigen | TUBGCP2 |
---|---|
Concentration | 300 μg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16864415
|
Proteintech
25856-1-AP-20UL |
20 μL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16854415
|
Proteintech
25856-1-AP-150UL |
150 μL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TUBGCP2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TUBGCP2. This antibody recognizes human antigen. The TUBGCP2 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
TUBGCP2 | |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG |
300 μg/mL | |
Polyclonal | |
Rabbit | |
10844 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title