Learn More
Masterflex™ Tygon™ 2 Stop Hydrocarbon Tubing
Tailored for petro-chemicals and resists swelling and cracking. Tygon™ 2 Stop Hydrocarbon Tubing is also UV and Ozone resistant.
590.00 DKK
Specifications
Material | Tygon F4040 |
---|---|
Overall Length (Metric) | 381 mm |
Packaging Quantity | 12 Pack |
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
11937865
|
Masterflex™
95606-22 |
0.64 mm |
590.00 DKK
Pack of 12 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
11907865
|
Masterflex™
95606-12 |
0.25 mm |
590.00 DKK
Pack of 12 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
11997865
|
Masterflex™
95606-34 |
1.42 mm |
590.00 DKK
Pack of 12 |
Estimated Shipment: 08-01-2026
Log in to see stock. |
|||||
11957865
|
Masterflex™
95606-26 |
0.89 mm |
590.00 DKK
Pack of 12 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
11967865
|
Masterflex™
95606-28 |
1.02 mm |
590.00 DKK
Pack of 12 |
Estimated Shipment: 20-10-2025
Log in to see stock. |
|||||
11987865
|
Masterflex™
95606-32 |
1.3 mm |
590.00 DKK
Pack of 12 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
|||||
11977865
|
Masterflex™
95606-30 |
1.14 mm |
590.00 DKK
Pack of 12 |
Estimated Shipment: 08-01-2026
Log in to see stock. |
|||||
11907875
|
Masterflex™
95606-36 |
1.5 mm |
590.00 DKK
Pack of 12 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
11927875
|
Masterflex™
95606-40 |
1.85 mm |
590.00 DKK
Pack of 12 |
Estimated Shipment: 20-10-2025
Log in to see stock. |
|||||
11937875
|
Masterflex™
95606-42 |
2.06 mm |
590.00 DKK
Pack of 12 |
Estimated Shipment: 06-02-2026
Log in to see stock. |
|||||
11947875
|
Masterflex™
95606-44 |
2.29 mm |
590.00 DKK
Pack of 12 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
Description
- Resists embrittlement
- Compatible with most hydrocarbons
- Resistant to swelling
- Highly flexible, easy to install
- Specifically developed for fuels and lubricants
- Ozone and UV light resistant
Specifications
Tygon F4040 | |
381 mm | |
12 Pack |