Learn More
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck
Delivers an ideal balance of protection, durability and comfort.
1955.00 DKK - 1990.00 DKK
Specifications
Product Type | Controlled Environments Lab Coat |
---|---|
Color | White |
Closure Type | Snaps |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16458363
|
DuPont™
D15535577 |
Small |
1955.00 DKK
Pack of 30 |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
16468363
|
DuPont™
D15535578 |
Medium |
1955.00 DKK
Pack of 30 |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
16478363
|
DuPont™
D15535579 |
1955.00 DKK
Pack of 30 |
Estimated Shipment: 04-09-2025
Log in to see stock. |
||||||
16488363
|
DuPont™
D15535580 |
X-Large |
1955.00 DKK
Pack of 30 |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
16498363
|
DuPont™
D15535581 |
2X-Large |
1955.00 DKK
Pack of 30 |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
16408373
|
DuPont™
D15535582 |
3X-Large |
1990.00 DKK
Pack of 30 |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
Description
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash
- Breathable and exceptionally low linting
Specifications
Controlled Environments Lab Coat | |
White | |
Snaps |