missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ Ultra Low Particulate Air (ULPA) Filter
GREENER_CHOICE

Ultra Low Particulate Air (ULPA) Filters are available in various configurations for the containment of particles.

8235.00 DKK - 9565.00 DKK

Specifications

Efficiency 99.9995%
Filter Type ULPA
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
17736004
GREENER_CHOICE
View Documents
Syngene™
SYN-ACFULPA18
No Frame
8235.00 DKK
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17746004
GREENER_CHOICE
View Documents
Syngene™
SYN-ACFULPA18W
Wood Frame
9565.00 DKK
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The filter has a 99.995% efficiency rating for collecting 0.125 micron particles.
Specifications

Specifications

99.9995%
ULPA
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.