Learn More
Nickel Electro™ Ultraclear Cleaning Solution
Nickel Electro™ Ultraclear Cleaning Solution is a general purpose cleaning agent.
Brand: Nickel Electro™ BX0613
296.37 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- For general purpose use
- Use with DU-4, DU-8, DU-14, DU-22, MU-8, MU-14, MU-22
Specifications
1 L | |
For MU-22, DU-22, DU-14, MU-14, MU-8, DU-8, DU-4 Ultrasonic Bath | |
Ultraclear Cleaning Solution |
Safety and Handling
- 1 LT Cleaning solution Ultraclear for ultrasonic
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.