Learn More
uvex Unidur 6659 Foam Cut Protection Glove
Gloves are constructed from a combination of HPPE and glass fibers to provide level C cut protection, with NBF foam coating on the palm and fingers for mechanical abrasion resistance.
97.85 DKK - 670.00 DKK
Specifications
Applications | Industrial |
---|---|
Coating Material | NBR Foam |
Color | Gray, Black |
Hand | Ambidextrous |
Latex Free | Yes |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17250481
|
uvex
6093806 |
6 |
97.85 DKK
1 pair |
In Stock
Log in to see stock. ![]() |
|||||
17260481
|
uvex
6093807 |
7 |
670.00 DKK
10 pairs |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
17270481
|
uvex
6093808 |
8 |
670.00 DKK
10 pairs |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
17280481
|
uvex
6093809 |
9 |
670.00 DKK
10 pairs |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
17290481
|
uvex
6093810 |
10 |
670.00 DKK
10 pairs |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
17200491
|
uvex
6093811 |
11 |
97.85 DKK
1 pair |
In Stock
Log in to see stock. ![]() |
|||||
Description
- HPPE and glass fiber construction provides level C cut protection
- NBR foam coating increases mechanical abrasion resistance
- Enhanced grip in dry or slightly damp areas
- Ideal for precision assembly work requiring tactility and sensitivity
Specifications
Industrial | |
Gray, Black | |
Yes | |
Cut-resistant Gloves | |
Automotive, Maintenance, Metal Work, and Shipping | |
Knit |
NBR Foam | |
Ambidextrous | |
HPPE, Fiberglass | |
4X43C | |
Unisex | |
Unidur 6659 Foam |