Learn More
Grant Instruments™ Adjustable Bars and Two Fixing Levels
Attach to universal platform to provide stability and security for vessels while orbital shaker is in motion. Grant Instruments™ Adjustable Bars and Two Fixing Levels do not require tools to attach to platform.
Brand: Grant Instruments™ PUP-330
4827.24 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- For use with Grant Orbital Shakers
- Dimensions: 345 x 430 x 105mm
- Includes Extra Holding Bars which can also be ordered separately (HB 330)
Specifications
345 x 430 x 105mm | |
PSU-2Oi |