Learn More
uvex Unipur 6639 Polyurethane-Coated Assembly Gloves
Unipur 6639 assembly gloves are suitable for precision mechanical work in dry to slightly damp work environments. The robust polyurethane coating provides abrasion-resistance, without compromising dexterity or tactility.
Brand: uvex 6024808
This item is not returnable.
View return policy
Description
- Polyamide gloves with polyurethane coating on the palm and fingertips
- Coating offers a high level of abrasion resistance
- Enhanced grip capabilities in dry, or slightly damp applications
- Lightweight and sensitive, ideal for precision mechanical work
- Dark colored material hides dirt and stains
Specifications
Polyurethane | |
Smooth | |
Polyamide | |
8 |
Black | |
Ambidextrous | |
Mechanical Protection Gloves | |
Full-hand Mitt |