Learn More
Thermo Scientific™ 96 Well Round (U) Bottom Plate, TC Surface, No Lid, Pack of 1
Color:Clear; Sterility:Sterile; Lid:Without Lid
Brand: Thermo Scientific™ 143761
788.74 DKK valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Description
Thermo Scientific™ Nunc MicroWell Plates with Nunclon Delta Surface are ideal in a wide variety of applications, including cloning experiments and tissue culture-based analytical systems. Thermo Scientific Nunc Cell Culture products offer solutions for cell growth and analysis and have been used by researchers worldwide since 1953. With a wide range of surface areas and treatments, we have products for all your basic or advanced cell-based research.
- Stackable
- Radiation sterilized
- Condensation rings control evaporation and retain condensate
- Keyed lids prevent accidental reversals
- Dimensions: 86 × 128mm
Ordering Information:
Individually wrapped.
Specifications
Custom Barcoding Available | |
Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility | |
8 x 12 | |
Nunc | |
Cell Culture Microplate | |
300 μL | |
Sterile | |
Round Bottom (U-well) | |
1 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Without Lid | |
10/Pack, 50/Case | |
Clear | |
Polystyrene | |
96 | |
Nunclon Delta | |
50 | |
Case of 50 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.