Learn More
Socorex™ Laboratory Pipette Racks
Hold and organize pipettes using Socorex™ Laboratory Pipette Stands. The space-saving work stations and shelf holder for pipettes come in a variety of colors. All are constructed of sturdy polyamid or polycarbonate materials and are very stable with anti-slip pads.
Brand:  Socorex™ 320.337R
 View more versions of this product
Product Code. 10266982
  • 933.00 DKK / Each
Only null left
Estimated Shipment: 01-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Especially suited for single- and multi-channel pipettes and macropipettes.
  • Accommodate all Calibra™ and Acura™ models
  • Available in pastel colors
  • Constructed of easy-to-clean polyamid and polycarbonate material
Specifications
Specifications
Universal Workstation
Model 337 Universal Work Station
Single-channel Micropipettes
7
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions