Learn More
SCAT™ Safety Waste Caps
GREENER_CHOICE
Safety Waste Caps for the safe disposal of liquid waste. For maximum protection and efficient work with hazardous waste liquids. Offers multiple options to connect different tubing and capillary sizes.
Brand:  SCAT™ 107257
1387.28 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12655895
  • 1715.00 DKK / Each
Only null left
Estimated Shipment: 17-06-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • 360° free turning PTFE body, resistant against aggressive media
  • Exhaust filter system against harmful vapors
  • Fittings and tube connectors in different sizes
  • Flexible system
  • Time saving in changing the waste containers while tubes are fixed
  • Lab safety, technical protective measures, optimal protection when collecting liquid waste
  • Cost efficiency
  • Huge range of accessories, optional: funnel use, level control preventing overflowing containers
  • Huge portfolio of thread sizes suitable for waste containers from different manufacturers
  • Safety disposal of HPLC waste
  • High work performance
  • Personal protection, avoid skin contact with organic solvents, occupational safety, ergonomics
  • Sealed bottles and containers
  • Create a comfortable working climate
Specifications
Specifications
Black
Waste Disposal Container
PTFE
Safety Waste Cap
HPLC Disposal Systems
4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter
S95
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions