Learn More
Brannan™ BS718 Density Hydrometers
Shot loaded density hydrometers for a variety of surface tension ranges.
Brand:  Brannan™ 10/336/0
 View more versions of this product
Product Code. 12811488
  • 149.00 DKK / Each
Only null left
Estimated Shipment: 29-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
Shot loaded density hydrometers for surface tension 0.700 to 0.750g/mL
  • Shot loaded
  • Length 190 mm
  • Scale subdivision 0.002
  • Accuracy ±0.002 g/mL
Specifications
Specifications
1 Ea.
0.001 g/mL
270 mm
0.90 to 0.95 g/mL
Density Hydrometer
UKAS
20°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions