Learn More
Cytiva Cytodex 3 Gamma Microcarriers
Shop All Cytiva Products
Click to view available options
Quantity:
30 g
300 g
3 kg
Unit Size:
300g
30g
3kg
Description
- Cytodex 3 Gamma is designed by chemically coupling of a thin layer of denatured porcine collagen type 1 to the cross-linked dextran matrix.
- As the collagen surface layer can be digested by a variety of proteolytic enzymes, harvest of such cells from the microcarriers can be performed with maintained high cell viability and membrane integrity.
- This feature of Cytodex 3 Gamma is beneficial in the development of successful serial subcultivation protocols for scaling up culture volumes.
- Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, differentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving cell transfer and studies on uptage of labelled compounds
Specifications
Specifications
For Use With (Application) | Production of large quantities of cells, viruses and recombinant cell products, studies on cell adhesion, differentiation and cell function |
Quantity | 30 g |
Safety and Handling
Hazard Statement
- EUH210-Safety data sheet available on request.
- UNKNOWN ACUTE-9.9 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction