Learn More
Memmert™ CO2 Incubator ICOmed (Medical Device)
Ideal for the creation and maintenance of constant environmental conditions for application in the field of in vitro fertilisation (IVF), especially for the incubation of oocytes, spermatozoa and zygotes in special culture dishes for IVF application.
Brand:  Memmert™ ICO240med
65043.13 DKK valid until 2025-12-31
Use promo code "25039" to get your promotional price.
 View more versions of this product
Product Code. 15861575
  • 85520.00 DKK / Each
Estimated Shipment: 13-08-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Sterile, safe and reliable – these three words describe the Memmert CO2 incubator ICOmed for the medical use.
Memmert subjected its CO2 incubator ICOmed to a comprehensive medical device evaluation. Each Memmert CO2 incubator ICOmed is classified as a Class IIa medical device for the in vitro fertilisation (IVF) as well as biosynthesis.
  • Model sizes: 50, 105, 150, 240
  • Temperature adjustment range: +18 °C to +50 °C
  • CO2 adjustment range: 0 to 20 %
  • Humidity adjustment ranges: 40 to 97 % rh (with option K7)
  • O2 adjustment ranges: 1 to 20 % (with option T6)
  • Classified as a medical device class IIa
  • 1 model variant: TwinDISPLAY (intuitive control)
  • Perfect distribution of temperature, humidity and CO2
  • Long-time stable and repetitively accurate
  • The interior chamber, including all installations and sensors, can be sterilised at +180 °C in a 60-minute programme
  • Visual and acoustic alarm as well as alarm notification to mobile phones if the individually adjustable temperature, CO2, O2, or humidity ranges are exceeded
  • Battery-buffered ControlCOCKPIT: The operating display, logging and CO2 control are fully functional even during a power failure
  • Fail-safe, FDA-compliant logging of temperature, CO2, O2 humidity, open door time data
  • Adjustable parameters: temperature (Celsius or Fahrenheit), CO2, programme time, time zones, summertime/wintertime
Active humidity control ensures short recovery times after the door has been opened and minimises vaporisation in the interior. Together with the heating of the interior from all six sides and the heated inner glass door, it offers maximum protection for cell and tissue cultures and avoids the dangerous formation of condensation. The turbulence-free chamber ventilation ensures a constant and uniform atmosphere.
Specifications
Specifications
Optic and acoustic
DIN 12880:2007-05 , EN 61010-1 (IEC 61010-1), EN 61010-2-010
241 L
0 to 20%
Textured Stainless Steel
1 to 20%
18°C to 50°C
60 min.
Mains cable with plug (German type)
500 x 600 x 810 mm
145 kg
115/230 V, 50/60 Hz
15 kg
TwinDISPLAY: adaptive multifunctional digital PID-microprocessor controller with 2 high-definition TFT-colour displays
Stainless Steel
IR Sensor
2
40 to 97%
CO2 Incubator
2 high-definition TFT-colour displays (TwinDISPLAY)
Perforated Stainless Steel
691 x 759 x 1176 mm
110 kg
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products