Learn More
Bel-Art™ Vikem Vinyl Aprons
Provides protection to personnel in hospitals, schools, laboratories and industrial plants.
320.00 DKK - 549.00 DKK
Specifications
Type | Apron |
---|---|
Size | Universal |
Material | Vinyl |
Color | Clear |
Disposable | Reusable |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10024610
|
Bel-Art™
H24603-0001 |
91 cm |
320.00 DKK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
|||||
10723461
|
Bel-Art™
H24604-0001 |
107 cm |
549.00 DKK
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
Description
- Lightweight and comfortable
- Made from 4mil thick vinyl
- Stitched cotton binding securing all edges
- Strong tie backs double reinforced with metal grommets through three layers of material at waist
- Non-absorbent and chemically resistant
Specifications
Apron | |
Vinyl | |
Reusable | |
Non-sterile | |
Sewn |
Universal | |
Clear | |
Ties | |
Stitched cotton binding securing all edges |