Learn More
Mutexil™ Non-woven Blue High-filter Visitors Face Masks
Click to view available options
Color:
Blue
Unit Size:
Pack of 1000
Description
- Non-woven
- Blue
- Easily fastened
- High filter for enhanced safety
Specifications
Specifications
Product Type | Face Mask |
Color | Blue |
Construction | Non-woven |
Filter Efficiency | High |
For Use With (Application) | For used to visit a manufacture or to visit patients |