Learn More
Thermo Scientific™ WaterBugs™ Enterobacter cloacae subsp. cloacae ATCC™ 13047™
Click to view available options
Quantity:
10 Tests/Kit
Unit Size:
Each
Description
Thermo Scientific™ WaterBugs™ Enterobacter cloacae subsp. cloacae ATCC™ 13047™ is used in water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
- Deliver <50 CFUs in a single inoculum to mimic water samples
- One test/reconstituted vial
Ordering Alerts:
Supplied with 10 preserved organism vials; 10 rehydration vials.
Specifications
Specifications
ATCC® | 13047TM*, 13047™* |
Description | Enterobacter cloacae subsp. cloacae ATCC™ 13047™ |
Format | Quanti-Cult |
Product Type | Quality Control Organism |
Target Organism Class | Gram-negative Rods |
Test Type | Quantitative QC |
Packaging Type | 10 Test/Kit |
Quantity | 10 Tests/Kit |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.