Learn More
DWK Life Sciences Wheaton™ BOD Bottles Replacement Robotic Stopper
Compatible with 60 and 300mL BOD bottles
Brand: DWK Life Sciences 227672
448.86 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Glass, closure-style stopper
- Unlined
Specifications
For 60 and 300mL BOD bottle | |
Glass | |
Pennyhead |