Learn More
DWK Life Sciences Wheaton™ Bottles Amber Wide-Mouth Packers with Polyvinyl-lined Closures
Amber glass for storing light-sensitive powders. Wheaton™ Bottles Amber Wide-Mouth Packers with Polyvinyl-lined Closures are available in several capacities.
255.00 DKK - 4195.00 DKK
Specifications
Color | Amber |
---|---|
Material | Glass |
Closure Color | White |
Closure Liner | Poly/Vinyl |
Closure Material | Polypropylene |
Product Code | Brand | Capacity (Metric) | Closure Size | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Quantity | Price | Quantity & Availability | |||
12349859
|
DWK Life Sciences
W216954 |
30 mL | 28-400 mm | 432/Cs. |
4195.00 DKK
Pack of 432 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
|||
11784189
|
DWK Life Sciences
W216939 |
30 mL | 28-400 mm | 24/Cs. |
255.00 DKK
Pack of 24 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
|||
12359859
|
DWK Life Sciences
W216955 |
60 mL | 33-400 mm | 216/Cs. |
2225.00 DKK
Pack of 216 |
|
|||
11774189
|
DWK Life Sciences
W216940 |
60 mL | 33-400 mm | 24/Cs. |
288.00 DKK
Pack of 24 |
In Stock
Log in to see stock. |
|||
12369859
|
DWK Life Sciences
W216956 |
125 mL | 38-400 mm | 180/Cs. |
2540.00 DKK
Pack of 180 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
|||
10689895
|
DWK Life Sciences
W216941 |
125 mL | 38-400 mm | 24/Cs. |
332.00 DKK
Pack of 24 |
In Stock
Log in to see stock. |
|||
11714199
|
DWK Life Sciences
W216942 |
250 mL | 45-400 mm | 24/Cs. |
421.00 DKK
Pack of 24 |
In Stock
Log in to see stock. |
|||
12379859
|
DWK Life Sciences
W216957 |
250 mL | 45-400 mm | 84/Cs. |
1460.00 DKK
Pack of 84 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
|||
12389859
|
DWK Life Sciences
W216958 |
500 mL | 53-400 mm | 60/Cs. |
1360.00 DKK
Pack of 60 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11714219
|
DWK Life Sciences
W216943 |
500 mL | 53-400 mm | 12/Cs. |
269.00 DKK
Pack of 12 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
|||
11759665
|
DWK Life Sciences
W216944 |
1000 mL | 53-400 mm | 12/Cs. |
372.00 DKK
Pack of 12 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
|||
12399859
|
DWK Life Sciences
W216959 |
1000 mL | 53-400 mm | 36/Cs. |
1175.00 DKK
Pack of 36 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
|||
Description
Amber glass for storing light-sensitive powders. Wheaton™ Bottles Amber Wide-Mouth Packers with Polyvinyl-lined Closures are available in several capacities.
- USP Type III soda-lime glass
- Allows for easy removal of contents
Specifications
Amber | |
White | |
Polypropylene |
Glass | |
Poly/Vinyl | |
Screw Cap |