Learn More
Euro Protection™ White SPP Polypropylene Lab Coat
White SPP Polypropylene Lab Coats with pockets
125.00€ - 223.00€
Specifications
Product Type | General Purpose Lab Coat |
---|---|
Gender | Unisex |
Disposable | Reusable |
Sterility | Non-sterile |
For Use With (Application) | Food, Industrial, Agriculture, Cleaning, Visitors |
Product Code | Brand | Size | Collar Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Collar Type | Price | Quantity & Availability | ||||
12003216
|
Euro Protection™
41113 |
Large | Classic |
158.00€
Pack of 50 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
12033216
|
Euro Protection™
41123 |
Large | Officer |
223.00€
Pack of 50 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
12013216
|
Euro Protection™
41114 |
X-Large | Classic |
125.00€
Pack of 50 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
Description
- Elasticated wrist
- For use in the food industry, cleaning work, etc.
Specifications
General Purpose Lab Coat | |
Reusable | |
Food, Industrial, Agriculture, Cleaning, Visitors | |
40 g/m2 |
Unisex | |
Non-sterile | |
2 Pockets |