Learn More
Buerkle™ Wide-Mouth Containers, HDPE
The Wide-mouth containers, HDPE are for storage or transport of liquids, powders or granules. The containers are made of HDPE.
312.46 DKK - 435.18 DKK
Specifications
Color | Translucent |
---|---|
Description | Wide mouth container |
For Use With (Application) | Storage or Transport of Liquids, Powders or Granules |
Includes | Screw cap |
Material | HDPE |
Alert:
Keep the container upright when transporting liquids!
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10798164
|
Buerkle™
1406-1005 |
5 L |
312.46 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10043872
|
Buerkle™
1406-1010 |
10 L |
435.18 DKK
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
Description
- Storage container
- Made of HDPE
- For liquids, powders or granules
Specifications
Translucent | |
Storage or Transport of Liquids, Powders or Granules | |
HDPE | |
Yes | |
Yes | |
Square | |
HDPE |
Wide mouth container | |
Screw cap | |
Each | |
Transparent | |
Wide | |
Wide Mouth Container | |
Screw Cap |