Learn More
Klever X-Change Plus (Wider Cut Head)
Recessed blade of X-Change Plus (Wider Cut Head) reduces cut injuries and damaged goods.
875.00 DKK
Specifications
Blade Material | Plastic Polymer |
---|---|
For Use With (Application) | Cuts Boxes, Film, Tape, and Thousands More Applications |
Disposable | Reusable |
Sterility | Non-sterile |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
17779017
|
Klever
PLS-200XC-30G |
Orange |
875.00 DKK
Pack of 50 |
Estimated Shipment: 30-09-2025
Log in to see stock. |
|||||
17789017
|
Klever
PLS-200XC-30W |
White |
875.00 DKK
Pack of 50 |
Estimated Shipment: 30-09-2025
Log in to see stock. |
|||||
17799017
|
Klever
PLS-200XC-30Y |
Yellow |
875.00 DKK
Pack of 50 |
Estimated Shipment: 30-09-2025
Log in to see stock. |
|||||
Description
- Cuts boxes, film, tape and thousands more applications.
- Lighter weight, multiple-position handle for control and reach Innovative tape splitter.
- Adavnced plastic polymers.
- Highest safety level.
- Interchangeable, replaceable head.
Specifications
Plastic Polymer | |
Cuts Boxes, Film, Tape, and Thousands More Applications | |
Reusable | |
Non-sterile |