Learn More
Meditrade™ Nitril™ 3000 Nitrile Examination Gloves
Non-sterile Meditrade™ disposable nitrile examination gloves provide excellent protection for environments with a high risk of contamination. Powder-free and latex-free they are ideal for use in clinical, laboratory, and manufacturing environments.
15.00 DKK - 19.00 DKK
Specifications
Certifications/Compliance AQL 1.5; EN ISO 13485:2016; 93/42/EWG / EEC: Class 1; PPE Category III; Food Compliance acc. to EC Directive 1935/2004 (blue only)
Description Single use disposable white nitrile gloves
Color White
Disposable Single-use
Latex Free Yes
View More Specs
Products 2
Product Code Brand Size Color Price Quantity & Availability  
Product Code Brand Size Color Price Quantity & Availability  
16693612
View Documents
Meditrade™
1280M
Medium White This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16682217
View Documents
Meditrade™
1280XL
X-Large White
19.00 DKK
Pack of 100
Only null left
Estimated Shipment: 04-08-2025
Add to basket
Description
Description
  • Examination glove, made of nitrile according to DIN EN 455
  • Protective glove according to PSA
  • Skin-friendly: powder-, protein- and latex-free
  • Micro-roughened for better grip
  • Elastic and tear resistant
  • Ambidextrous fit, with rolled edge
  • Suitable for contact with food
Applications:Suitable for handling cleaning agents and disinfection and for other applications in the laboratory, pharmaceutical industry, professional cleaning, catering and food handling in general.
Specifications
Specifications
AQL 1.5; EN ISO 13485:2016; 93/42/EWG / EEC: Class 1; PPE Category III; Food Compliance acc. to EC Directive 1935/2004 (blue only)
White
Yes
0.060 mm (Cuff)
Single use disposable white nitrile gloves
Single-use
Nitrile